siRNA Details
| Gene Symbol: Cfhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044227.1 | NP_001037692.1 | 2 | 280 | ||
| XM_006249941.1 | XP_006250003.1 | 3 | 391 | |||
| GenBank mRNA | BC088173.1 | - | 2 | 280 | 7323337 | |
| FQ218705.1 | - | 2 | 298 | |||
| FQ219461.1 | - | 2 | 301 | |||
| Gene Symbol: LOC102554977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249952.2 | XP_006250014.2 | 2 | 315 | ||

Cfhr1
Genomic Map