siRNA Details
| Gene Symbol: Ivns1abp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047085.1 | NP_001040550.1 | 5 | 664 | ||
| XM_006249988.2 | XP_006250050.1 | 10 | 1277 | |||
| GenBank mRNA | AY553870.2 | - | 5 | 664 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ivns1abp| Gene Symbol: Ivns1abp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047085.1 | NP_001040550.1 | 5 | 664 | ||
| XM_006249988.2 | XP_006250050.1 | 10 | 1277 | |||
| GenBank mRNA | AY553870.2 | - | 5 | 664 | ||