siRNA Details
| Gene Symbol: Nmnat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048042.1 | NP_001041507.1 | 5 | 508 | ||
| XM_008769614.1 | XP_008767836.1 | 5 | 366 | |||
| GenBank mRNA | DQ022370.1 | - | 5 | 508 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nmnat2| Gene Symbol: Nmnat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048042.1 | NP_001041507.1 | 5 | 508 | ||
| XM_008769614.1 | XP_008767836.1 | 5 | 366 | |||
| GenBank mRNA | DQ022370.1 | - | 5 | 508 | ||