siRNA Details
| Gene Symbol: Acbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011906.1 | NP_001011906.1 | 1, 2 | 731 | ||
| XM_006250031.2 | XP_006250093.1 | 1, 2 | 787 | |||
| XM_006250032.2 | XP_006250094.1 | 1, 2 | 795 | |||
| XM_006250033.2 | XP_006250095.1 | 1, 2 | 794 | |||
| XM_006250035.2 | XP_006250097.1 | 1, 2 | 788 | |||
| XM_006250036.2 | XP_006250098.1 | 1, 2 | 788 | |||
| GenBank mRNA | BC086598.1 | - | 1, 2 | 731 | 7313105 | |

Acbd6
Genomic Map