siRNA Details
| Gene Symbol: Acbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011906.1 | NP_001011906.1 | 5 | 990 | ||
| GenBank mRNA | BC086598.1 | - | 5 | 990 | 7313105 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acbd6| Gene Symbol: Acbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011906.1 | NP_001011906.1 | 5 | 990 | ||
| GenBank mRNA | BC086598.1 | - | 5 | 990 | 7313105 | |