siRNA Details
| Gene Symbol: Tp53i3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113775.1 | NP_001107247.1 | 5 | 391 | ||
| XM_006250040.2 | XP_006250102.1 | 6 | 1174 | |||
| XM_008769617.1 | XP_008767839.1 | 5 | 1259 | |||
| XM_008769618.1 | XP_008767840.1 | 3 | 1102 | |||
| XM_008769619.1 | XP_008767841.1 | 5 | 422 | |||
| XM_008769620.1 | XP_008767842.1 | 5 | 338 | |||
| XM_008769621.1 | XP_008767843.1 | 2 | 1037 | |||
| GenBank mRNA | BC158771.1 | - | 6 | 438 | 7124147 | |

Tp53i3
Genomic Map