siRNA Details
| Gene Symbol: Mrps14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105963.2 | NP_001099433.1 | 3 | 340 | ||
| XM_006250098.2 | XP_006250160.1 | 3 | 457 | |||
| GenBank mRNA | BC166895.1 | - | 3 | 340 | 6920474 | |
| FQ214536.1 | - | 3 | 324 | |||

Mrps14
Genomic Map