siRNA Details
| Gene Symbol: Kifap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105964.1 | NP_001099434.1 | 19 | 2503 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kifap3| Gene Symbol: Kifap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105964.1 | NP_001099434.1 | 19 | 2503 | ||