siRNA Details
| Gene Symbol: Slc19a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030024.1 | NP_001025195.1 | 1, 2 | 365 | ||
| GenBank mRNA | BC099811.1 | - | 1, 2 | 365 | 7929969 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc19a2| Gene Symbol: Slc19a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030024.1 | NP_001025195.1 | 1, 2 | 365 | ||
| GenBank mRNA | BC099811.1 | - | 1, 2 | 365 | 7929969 | |