siRNA Details
| Gene Symbol: Creg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105966.1 | NP_001099436.1 | 3 | 542 | ||
| XM_008769625.1 | XP_008767847.1 | 3 | 666 | |||
| GenBank mRNA | BC166446.1 | - | 3 | 556 | 8364240 | |
| FQ227495.1 | - | 3 | 546 | |||
| FQ230214.1 | - | 3 | 575 | |||
| FQ230343.1 | - | 3 | 540 | |||
| FQ231443.1 | - | 3 | 529 | |||
| FQ232342.1 | - | 3 | 546 | |||
| FQ234634.1 | - | 3 | 575 | |||

Creg1
Genomic Map