siRNA Details
| Gene Symbol: Nhlh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105970.1 | NP_001099440.1 | 2 | 507 | ||
| XM_006250280.1 | XP_006250342.1 | 3 | 599 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nhlh1| Gene Symbol: Nhlh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105970.1 | NP_001099440.1 | 2 | 507 | ||
| XM_006250280.1 | XP_006250342.1 | 3 | 599 | |||