siRNA Details
| Gene Symbol: Olr1601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000087.1 | NP_001000087.1 | 1 | 870 | ||
| Gene Symbol: LOC103693685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769806.1 | XP_008768028.1 | 870 | |||

Olr1601
Genomic Map