siRNA Details
| Gene Symbol: Desi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013873.1 | NP_001013895.1 | 4, 5 | 759 | ||
| XM_006250316.2 | XP_006250378.1 | 4, 5 | 1109 | |||
| XM_006250317.2 | XP_006250379.1 | 5, 6 | 1523 | |||
| XM_008769783.1 | XP_008768005.1 | 4, 5 | 1066 | |||
| XM_008769784.1 | XP_008768006.1 | 5, 6 | 1242 | |||
| GenBank mRNA | BC083584.1 | - | 4, 5 | 759 | 7131107 | |
| FQ226068.1 | - | 4, 5 | 534 | |||

Desi2
Genomic Map