siRNA Details
| Gene Symbol: Efcab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105977.1 | NP_001099447.1 | 3 | 361 | ||
| XM_006250319.2 | XP_006250381.1 | 4 | 689 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efcab2| Gene Symbol: Efcab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105977.1 | NP_001099447.1 | 3 | 361 | ||
| XM_006250319.2 | XP_006250381.1 | 4 | 689 | |||