siRNA Details
| Gene Symbol: Gpr137b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105978.1 | NP_001099448.1 | 4 | 526 | ||
| XM_006254372.2 | XP_006254434.1 | 5 | 1028 | |||
| XM_006254373.2 | XP_006254435.1 | 5 | 1023 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr137b| Gene Symbol: Gpr137b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105978.1 | NP_001099448.1 | 4 | 526 | ||
| XM_006254372.2 | XP_006254434.1 | 5 | 1028 | |||
| XM_006254373.2 | XP_006254435.1 | 5 | 1023 | |||