siRNA Details
| Gene Symbol: Mixl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105979.1 | NP_001099449.1 | 1 | 40 | ||
| XM_008769818.1 | XP_008768040.1 | 1 | 18 | |||
| Gene Symbol: LOC100912275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751285.3 | XP_003751333.1 | 1 | 18 | ||
| XM_008769810.1 | XP_008768032.1 | 1 | 18 | |||

Mixl1
Genomic Map