siRNA Details
| Gene Symbol: Sde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024970.2 | NP_001020141.2 | 7 | 1381 | ||
| GenBank mRNA | BC091401.1 | - | 4 | 1264 | 7324146 | |
| CV077309.1 | - | 2 | 513 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sde2| Gene Symbol: Sde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024970.2 | NP_001020141.2 | 7 | 1381 | ||
| GenBank mRNA | BC091401.1 | - | 4 | 1264 | 7324146 | |
| CV077309.1 | - | 2 | 513 | |||