siRNA Details
| Gene Symbol: Sde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024970.2 | NP_001020141.2 | 5 | 629 | ||
| GenBank mRNA | BC091401.1 | - | 2 | 512 | 7324146 | |
| CB724808.1 | - | 2 | 84 | |||
| DY308690.1 | - | 5 | 629 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sde2| Gene Symbol: Sde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024970.2 | NP_001020141.2 | 5 | 629 | ||
| GenBank mRNA | BC091401.1 | - | 2 | 512 | 7324146 | |
| CB724808.1 | - | 2 | 84 | |||
| DY308690.1 | - | 5 | 629 | |||