siRNA Details
| Gene Symbol: Lefty2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007556.1 | NP_001007557.1 | 1 | 82 | ||
| GenBank mRNA | AY758558.1 | - | 1 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lefty2| Gene Symbol: Lefty2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007556.1 | NP_001007557.1 | 1 | 82 | ||
| GenBank mRNA | AY758558.1 | - | 1 | 82 | ||