siRNA Details
| Gene Symbol: Tmem63a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134496.2 | NP_001127968.2 | 11, 12 | 1054 | ||
| XM_006250364.2 | XP_006250426.1 | 11, 12 | 1231 | |||
| XM_006250365.2 | XP_006250427.1 | 11, 12 | 1086 | |||
| GenBank mRNA | FQ210846.1 | - | 11, 12 | 1026 | ||

Tmem63a
Genomic Map