siRNA Details
| Gene Symbol: Susd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105982.1 | NP_001099452.1 | 6 | 979 | ||
| XM_006250373.2 | XP_006250435.1 | 6 | 1139 | |||
| XM_006250375.2 | XP_006250437.1 | 6 | 1018 | |||
| XM_008769820.1 | XP_008768042.1 | 6 | 1139 | |||
| XM_008769821.1 | XP_008768043.1 | 6 | 1028 | |||
| XM_008769822.1 | XP_008768044.1 | 6 | 1117 | |||
| XM_008769823.1 | XP_008768045.1 | 6 | 1133 | |||
| GenBank mRNA | FQ212123.1 | - | 6 | 963 | ||

Susd4
Genomic Map