siRNA Details
| Gene Symbol: Rps6kc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105984.1 | NP_001099454.1 | 10 | 1490 | ||
| XM_006250459.2 | XP_006250521.1 | 11 | 1669 | |||
| XM_006250460.2 | XP_006250522.1 | 10 | 1576 | |||
| XM_006250461.2 | XP_006250523.1 | 11 | 1808 | |||
| XM_008769829.1 | XP_008768051.1 | 11 | 1669 | |||
| XM_008769830.1 | XP_008768052.1 | 10 | 1309 | |||
| XM_008769831.1 | XP_008768053.1 | 10 | 1446 | |||
| XM_008769832.1 | XP_008768054.1 | 4 | 4020 | |||
| XM_008769833.1 | XP_008768055.1 | 4 | 705 | |||
| XM_008769834.1 | XP_008768056.1 | 5 | 639 | |||
| GenBank mRNA | BC064438.1 | - | 11 | 1740 | ||

Rps6kc1
Genomic Map