siRNA Details
| Gene Symbol: Rps6kc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105984.1 | NP_001099454.1 | 6 | 897 | ||
| XM_006250459.2 | XP_006250521.1 | 6 | 953 | |||
| XM_006250460.2 | XP_006250522.1 | 6 | 953 | |||
| XM_006250461.2 | XP_006250523.1 | 6 | 1092 | |||
| XM_008769829.1 | XP_008768051.1 | 6 | 953 | |||
| XM_008769831.1 | XP_008768053.1 | 5 | 730 | |||
| GenBank mRNA | BC064438.1 | - | 6 | 1024 | ||

Rps6kc1
Genomic Map