siRNA Details
| Gene Symbol: Rab3gap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040154.2 | NP_001035244.2 | 34 | 4062 | ||
| XM_008769835.1 | 35 | 4047 | ||||
| XM_008769836.1 | XP_008768058.1 | 33 | 5468 | |||
| XM_008769837.1 | XP_008768059.1 | 32 | 3738 | |||
| GenBank mRNA | BC086380.1 | - | 8 | 903 | ||

Rab3gap2
Genomic Map