siRNA Details
| Gene Symbol: Gpatch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011909.1 | NP_001011909.1 | 4 | 1003 | ||
| XM_008769838.1 | XP_008768060.1 | 4 | 1002 | |||
| XM_008769839.1 | XP_008768061.1 | 4 | 3652 | |||
| XM_008769840.1 | XP_008768062.1 | 4 | 1087 | |||
| XM_008769841.1 | XP_008768063.1 | 5 | 1193 | |||
| XM_008769842.1 | XP_008768064.1 | 4 | 1002 | |||
| XM_008769843.1 | XP_008768065.1 | 4 | 1002 | |||
| XM_008769844.1 | XP_008768066.1 | 4 | 1002 | |||
| XM_008769845.1 | XP_008768067.1 | 4 | 1002 | |||
| GenBank mRNA | BC079232.1 | - | 4 | 1003 | 7132530 | |

Gpatch2
Genomic Map