siRNA Details
| Gene Symbol: Nenf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002851.1 | NP_001002851.1 | 3 | 312 | ||
| GenBank mRNA | AY623793.1 | - | 3 | 312 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nenf| Gene Symbol: Nenf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002851.1 | NP_001002851.1 | 3 | 312 | ||
| GenBank mRNA | AY623793.1 | - | 3 | 312 | ||