siRNA Details
| Gene Symbol: Atp6v1g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105991.1 | NP_001099461.1 | 3 | 285 | ||
| XM_006249924.2 | XP_006249986.1 | 3 | 1195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v1g3| Gene Symbol: Atp6v1g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105991.1 | NP_001099461.1 | 3 | 285 | ||
| XM_006249924.2 | XP_006249986.1 | 3 | 1195 | |||