siRNA Details
| Gene Symbol: Xpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105992.1 | NP_001099462.1 | 10 | 1380 | ||
| XM_008769626.1 | XP_008767848.1 | 10 | 2269 | |||
More Information
Other Species Targeted
Set Membership: |
|

Xpr1
Genomic Map