siRNA Details
| Gene Symbol: Glmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105993.1 | NP_001099463.1 | 15, 16 | 1521 | ||
| XM_008769921.1 | XP_008768143.1 | 14, 15 | 2128 | |||
| XM_008769922.1 | XP_008768144.1 | 15, 16 | 1522 | |||
| XM_008769923.1 | XP_008768145.1 | 15, 16 | 1601 | |||
| XM_008769924.1 | XP_008768146.1 | 14, 15 | 2128 | |||
| XM_008769925.1 | XP_008768147.1 | 13, 14 | 2187 | |||
| XM_008769926.1 | XP_008768148.1 | 13, 14 | 1314 | |||
| GenBank mRNA | BC083633.1 | - | 13, 14 | 1543 | ||
| BC100158.1 | - | 15, 16 | 1634 | |||
| BC162027.1 | - | 15, 16 | 1497 | |||

Glmn
Genomic Map