siRNA Details
| Gene Symbol: Ugt2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135869.1 | NP_001129341.1 | 169 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugt2a3| Gene Symbol: Ugt2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135869.1 | NP_001129341.1 | 169 | |||