siRNA Details
Gene Symbol: Tmprss11g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008554.1 | NP_001008554.1 | 6 | 565 | ||
GenBank mRNA | AJ617528.1 | - | 6 | 565 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmprss11g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008554.1 | NP_001008554.1 | 6 | 565 | ||
GenBank mRNA | AJ617528.1 | - | 6 | 565 |