siRNA Details
| Gene Symbol: Spata18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199374.2 | NP_955406.2 | 6 | 888 | ||
| XM_006250891.2 | XP_006250953.1 | 6 | 899 | |||
| GenBank mRNA | AB116526.1 | - | 6 | 838 | ||
| BC078996.1 | - | 6 | 888 | 7112725 | ||

Spata18
Genomic Map