siRNA Details
| Gene Symbol: Atp8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250974.2 | XP_006251036.1 | 3 | 478 | ||
| XM_006250975.2 | XP_006251037.1 | 3 | 478 | |||
| XM_006250976.2 | XP_006251038.1 | 3 | 478 | |||
| XM_006250977.2 | XP_006251039.1 | 3 | 478 | |||
| XM_008770164.1 | XP_008768386.1 | 3 | 478 | |||

Atp8a1
Genomic Map