siRNA Details
| Gene Symbol: Evc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106012.1 | NP_001099482.1 | 5 | 617 | ||
| XM_006251088.2 | XP_006251150.1 | 5 | 481 | |||
| XM_008770206.1 | XP_008768428.1 | 5 | 974 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Evc2| Gene Symbol: Evc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106012.1 | NP_001099482.1 | 5 | 617 | ||
| XM_006251088.2 | XP_006251150.1 | 5 | 481 | |||
| XM_008770206.1 | XP_008768428.1 | 5 | 974 | |||