siRNA Details
| Gene Symbol: Pes1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044228.1 | NP_001037693.1 | 2 | 101 | ||
| GenBank mRNA | BC105910.1 | - | 2 | 101 | 7388101 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pes1| Gene Symbol: Pes1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044228.1 | NP_001037693.1 | 2 | 101 | ||
| GenBank mRNA | BC105910.1 | - | 2 | 101 | 7388101 | |