siRNA Details
| Gene Symbol: Osm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006961.1 | NP_001006962.1 | 2 | 125 | ||
| GenBank mRNA | AB167521.1 | - | 2 | 125 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osm| Gene Symbol: Osm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006961.1 | NP_001006962.1 | 2 | 125 | ||
| GenBank mRNA | AB167521.1 | - | 2 | 125 | ||