siRNA Details
| Gene Symbol: LOC100912481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749683.2 | XP_003749731.1 | 14 | 1522 | ||
| XM_003749684.2 | XP_003749732.1 | 15 | 1537 | |||
| XM_006236016.1 | XP_006236078.1 | 15 | 1540 | |||
| XM_006236017.1 | XP_006236079.1 | 14 | 1519 | |||
| Gene Symbol: Ewsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025632.2 | NP_001020803.2 | 14 | 1517 | ||
| XM_006251201.2 | XP_006251263.2 | 14 | 1502 | |||
| XM_006251202.2 | XP_006251264.2 | 14 | 1499 | |||
| XM_006251203.2 | XP_006251265.2 | 14 | 1528 | |||
| GenBank mRNA | BC098822.1 | - | 5 | 551 | ||
| BC162004.1 | - | 14 | 1517 | 9089248 | ||
| FQ221459.1 | - | 5 | 664 | |||

LOC100912481
Genomic Map