siRNA Details
| Gene Symbol: Myl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106017.1 | NP_001099487.1 | 2, 3 | 205 | ||
| XM_006251216.2 | XP_006251278.1 | 3, 4 | 305 | |||
| GenBank mRNA | BC161995.1 | - | 205 | 9034230 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myl7| Gene Symbol: Myl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106017.1 | NP_001099487.1 | 2, 3 | 205 | ||
| XM_006251216.2 | XP_006251278.1 | 3, 4 | 305 | |||
| GenBank mRNA | BC161995.1 | - | 205 | 9034230 | ||