siRNA Details
| Gene Symbol: Ugp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024743.1 | NP_001019914.1 | 8 | 1365 | ||
| XM_006251538.1 | XP_006251600.1 | 8 | 1317 | |||
| GenBank mRNA | BC084711.1 | - | 3 | 472 | ||
| BC097369.1 | - | 8 | 1365 | 7443949 | ||
| FQ214781.1 | - | 3 | 548 | |||

Ugp2
Genomic Map