siRNA Details
Gene Symbol: Psmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100509.1 | NP_001093979.1 | 1 | 120 | ||
GenBank mRNA | BC107950.1 | - | 1 | 41 | ||
DY570284.1 | - | 1 | 120 | |||
FQ213949.1 | - | 1 | 68 | |||
FQ214043.1 | - | 1 | 73 | |||
FQ227742.1 | - | 1 | 77 |
