siRNA Details
| Gene Symbol: Cnih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106029.2 | NP_001099499.1 | 1, 2 | 112 | ||
| GenBank mRNA | BC158747.1 | - | 1, 2 | 112 | 9101224 | |
| FQ222749.1 | - | 1, 2 | 161 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnih1| Gene Symbol: Cnih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106029.2 | NP_001099499.1 | 1, 2 | 112 | ||
| GenBank mRNA | BC158747.1 | - | 1, 2 | 112 | 9101224 | |
| FQ222749.1 | - | 1, 2 | 161 | |||