siRNA Details
| Gene Symbol: Olr1610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000094.1 | NP_001000094.1 | 1 | 596 | ||
| XM_008770652.1 | XP_008768874.1 | 1 | 599 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1610| Gene Symbol: Olr1610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000094.1 | NP_001000094.1 | 1 | 596 | ||
| XM_008770652.1 | XP_008768874.1 | 1 | 599 | |||