siRNA Details
| Gene Symbol: Cmtm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106034.2 | NP_001099504.2 | 1 | 379 | ||
| GenBank mRNA | FM029909.1 | - | 1 | 379 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmtm5| Gene Symbol: Cmtm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106034.2 | NP_001099504.2 | 1 | 379 | ||
| GenBank mRNA | FM029909.1 | - | 1 | 379 | ||