siRNA Details
| Gene Symbol: RGD1564324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106035.1 | NP_001099505.1 | 1, 2 | 94 | ||
| XM_008770666.1 | XP_008768888.1 | 2, 3 | 335 | |||
| XM_008770671.1 | XP_008768893.1 | 2, 3 | 335 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564324| Gene Symbol: RGD1564324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106035.1 | NP_001099505.1 | 1, 2 | 94 | ||
| XM_008770666.1 | XP_008768888.1 | 2, 3 | 335 | |||
| XM_008770671.1 | XP_008768893.1 | 2, 3 | 335 | |||