siRNA Details
| Gene Symbol: Xpo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106042.1 | NP_001099512.1 | 19 | 3073 | ||
| XM_006252062.2 | XP_006252124.1 | 19 | 3077 | |||
| XM_006252063.2 | XP_006252125.1 | 19 | 2949 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xpo4| Gene Symbol: Xpo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106042.1 | NP_001099512.1 | 19 | 3073 | ||
| XM_006252062.2 | XP_006252124.1 | 19 | 3077 | |||
| XM_006252063.2 | XP_006252125.1 | 19 | 2949 | |||