siRNA Details
| Gene Symbol: Sap18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033685.2 | NP_001028857.2 | 3 | 309 | ||
| GenBank mRNA | BC104706.1 | - | 3 | 298 | 7316605 | |
| CK357541.1 | - | 3 | 266 | |||
| FQ212380.1 | - | 3 | 281 | |||
| FQ223271.1 | - | 3 | 276 | |||
| FQ234134.1 | - | 3 | 317 | |||
| Gene Symbol: RGD1561590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002727811.4 | XP_002727857.1 | 1 | 332 | ||

Sap18
Genomic Map