siRNA Details
Gene Symbol: Dleu7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106043.1 | NP_001099513.1 | 1 | 292 | ||
XM_006252176.2 | XP_006252238.1 | 1 | 623 | |||
GenBank mRNA | BC158600.1 | - | 1 | 140 | 7130710 |
