siRNA Details
| Gene Symbol: Dleu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106043.1 | NP_001099513.1 | 3 | 871 | ||
| XM_006252176.2 | XP_006252238.1 | 2 | 1062 | |||
| GenBank mRNA | BC158600.1 | - | 2 | 579 | 7130710 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dleu7| Gene Symbol: Dleu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106043.1 | NP_001099513.1 | 3 | 871 | ||
| XM_006252176.2 | XP_006252238.1 | 2 | 1062 | |||
| GenBank mRNA | BC158600.1 | - | 2 | 579 | 7130710 | |