siRNA Details
Gene Symbol: Dnajc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106050.1 | NP_001099520.1 | 4 | 440 | ||
GenBank mRNA | FQ229062.1 | - | 4 | 314 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dnajc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106050.1 | NP_001099520.1 | 4 | 440 | ||
GenBank mRNA | FQ229062.1 | - | 4 | 314 |