siRNA Details
| Gene Symbol: Dnajc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106050.1 | NP_001099520.1 | 5 | 476 | ||
| GenBank mRNA | FQ229062.1 | - | 5 | 350 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc15| Gene Symbol: Dnajc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106050.1 | NP_001099520.1 | 5 | 476 | ||
| GenBank mRNA | FQ229062.1 | - | 5 | 350 | ||